1-(2-Amino-5-fluorophenyl)ethanone 97+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC201208-5G
APOSPC201208-5GEA 46.2 GBP
APOSPC201208-5G
1-(2-Amino-5-fluorophenyl)ethanone 97+%
1-(2-Amino-5-fluorophenyl)ethanone
Formula: C₈H₈FNO
MW: 153.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835333
CAS Number: 2343-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR