Pentafluoroethyl methyl ether 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC1289-100G PC1289-25G
APOSPC1289-100GEA 823 GBP
APOSPC1289-100G APOSPC1289-25G
Pentafluoroethyl methyl ether 98%
Pentafluoroethyl methyl ether

  • Non-flammable gas

Formula: C₃H₃F₅O
Boiling Pt: 6 °C/738mmHg
Melting Pt: –115 °C
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094117
CAS Number: 22410-44-2
UN: 3163
ADR: 2.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR