4-Fluoro-3-(tributylstannyl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC1191-1G PC1191-250MG
APOSPC1191-1GEA 237 GBP
APOSPC1191-1G APOSPC1191-250MG
4-Fluoro-3-(tributylstannyl)pyridine
4-Fluoro-3-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
Storage Temperature: Ambient
CAS Number: 918542-03-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR