3-Fluoro-2-(tributylstannyl)pyridine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC1189-1G PC1189-5G
APOSPC1189-1GEA 166 GBP
APOSPC1189-1G APOSPC1189-5G
3-Fluoro-2-(tributylstannyl)pyridine 98%
3-Fluoro-2-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
Storage Temperature: Refrigerator
CAS Number: 573675-60-2
UN: 2788
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR