2-Fluoro-6-hydroxybenzonitrile 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC10435-1G PC10435-25G PC10435-5G
APOSPC10435-1GEA 10.6 GBP
APOSPC10435-1G APOSPC10435-25G APOSPC10435-5G
2-Fluoro-6-hydroxybenzonitrile 99%
2-Fluoro-6-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137.11 g/mol
Melting Pt: 154…158 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03428592
CAS Number: 140675-43-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR