3-(Methylamino)benzotrifluoride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC10433-1G PC10433-25G PC10433-5G
APOSPC10433-1GEA 29.6 GBP
APOSPC10433-1G APOSPC10433-25G APOSPC10433-5G
3-(Methylamino)benzotrifluoride
3-(Methylamino)benzotrifluoride
Formula: C₈H₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD07368696
CAS Number: 2026-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR