1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC1043-1G PC1043-250MG
APOSPC1043-1GEA 581 GBP
APOSPC1043-1G APOSPC1043-250MG
1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
Formula: C₁₇H₃₁F₃N₂Sn
Storage Temperature: Ambient
MDL Number: MFCD06797466
CAS Number: 191606-78-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR