1,1,1,3,3,3-Hexafluoro-2-propanol 99,9%, Spectrophotometric Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

PC0877-100ML PC0877-25ML PC0877-500ML
APOSPC0877-100MLEA 117 GBP
APOSPC0877-100ML APOSPC0877-25ML APOSPC0877-500ML
1,1,1,3,3,3-Hexafluoro-2-propanol 99,9%, Spectrophotometric Grade
1,1,1,3,3,3-Hexafluoro-2-propanol

UV absorption < 0.1nm between 200 - 400 nm

Formula: C₃H₂F₆O
Boiling Pt: 58 °C
Melting Pt: –4 °C
Density: 1.596 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR