2-Chloro-3,3,3-trifluoropropene 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC0682-100G PC0682-25G
APOSPC0682-100GEA 345 GBP
APOSPC0682-100G APOSPC0682-25G
2-Chloro-3,3,3-trifluoropropene 98%
2-Chloro-3,3,3-trifluoropropene
Formula: C₃H₂ClF₃
Boiling Pt: 14…15 °C
Storage Temperature: Ambient
MDL Number: MFCD00082613
CAS Number: 2730-62-3
UN: 3163
ADR: 2.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR