Hexafluoroisopropyl methyl ether 99+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl hexafluoroisopropyl ether

PC0362-25G PC0362-5G
APOSPC0362-25GEA 458 GBP
APOSPC0362-25G APOSPC0362-5G
Hexafluoroisopropyl methyl ether 99+%
Hexafluoroisopropyl methyl ether
Formula: C₄H₄F₆O
MW: 182.064 g/mol
Boiling Pt: 29 °C
Density: 1.42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00221772
CAS Number: 22052-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR