Perfluorohexane 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tetradecafluorohexane

PC0098-25G PC0098-100G
APOSPC0098-25GEA 73.9 GBP
APOSPC0098-25G APOSPC0098-100G
Perfluorohexane 99%
Perfluorohexane
Formula: C₆F₁₄
Boiling Pt: 59…60 °C
Density: 1.68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000437
CAS Number: 355-42-0
EINECS: 206-585-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR