5-Bromovaleric acid 97+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR9972-100G OR9972-25G
APOSOR9972-100GEA 68.3 GBP
APOSOR9972-100G APOSOR9972-25G
5-Bromovaleric acid 97+%
5-Bromovaleric acid
Formula: BrH₂C(CH₂)₃COOH
Boiling Pt: 142…145 °C/13mmHg
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004414
CAS Number: 2067-33-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR