4-Iodotetrahydropyran 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Iodotetrahydro-2H-pyran

OR9693-1G OR9693-5G
APOSOR9693-1GEA 15.8 GBP
APOSOR9693-1G APOSOR9693-5G
4-Iodotetrahydropyran 97%
4-Iodotetrahydropyran
Formula: C₅H₉IO
Boiling Pt: 79…80 °C/12mmHg
Storage Temperature: Ambient
MDL Number: MFCD06797467
CAS Number: 25637-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR