3-Isobutoxybenzoic acid 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR953952-10G
OR953952-1G
OR953952-5G
APOSOR953952-10GEA
494
GBP
APOSOR953952-10G
APOSOR953952-1G
APOSOR953952-5G
3-Isobutoxybenzoic acid 98%
3-Isobutoxybenzoic acid
Formula:
C₁₁H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD01993662 CAS Number: 350997-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...