Ethyl-4-acetylbenzoate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR953424-1G OR953424-5G
APOSOR953424-1GEA 17.7 GBP
APOSOR953424-1G APOSOR953424-5G
Ethyl-4-acetylbenzoate 95%
Ethyl-4-acetylbenzoate
Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00013241
CAS Number: 38430-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR