2-Isopropoxybenzaldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR946443-10G OR946443-5G
APOSOR946443-10GEA 0 GBP
APOSOR946443-10G APOSOR946443-5G
2-Isopropoxybenzaldehyde 98%
2-Isopropoxybenzaldehyde
Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01571834
CAS Number: 22921-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR