5-tert-Butyl-1,3,4-oxadiazol-2-amine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR943775-1G OR943775-250MG
APOSOR943775-1GEA 115 GBP
APOSOR943775-1G APOSOR943775-250MG
5-tert-Butyl-1,3,4-oxadiazol-2-amine 95%
5-tert-Butyl-1,3,4-oxadiazol-2-amine
Formula: C₆H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD09261284
CAS Number: 69741-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR