1-Acetyl-2-piperidinecarboxylic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR943768-1G OR943768-500MG
APOSOR943768-1GEA 286 GBP
APOSOR943768-1G APOSOR943768-500MG
1-Acetyl-2-piperidinecarboxylic acid 97%
1-Acetyl-2-piperidinecarboxylic acid
Formula: C₈H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD09044120
CAS Number: 35677-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR