N-Vinylacetamide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR938950-25G OR938950-5G
APOSOR938950-25GEA 185 GBP
APOSOR938950-25G APOSOR938950-5G
N-Vinylacetamide 98%
N-Vinylacetamide
Formula: C₄H₇NO
Storage Temperature: Ambient
MDL Number: MFCD00081205
CAS Number: 5202-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR