Ethoxyquin 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

OR938378-10G OR938378-100G OR938378-1KG
APOSOR938378-10GEA 15.9 GBP
APOSOR938378-10G APOSOR938378-100G APOSOR938378-1KG
Ethoxyquin 95%
Ethoxyquin
Formula: C₁₄H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD00023883
CAS Number: 91-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR