O-Benzoyl-N-methylhydroxylamine hydrochloride 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR937790-10G
OR937790-1G
OR937790-5G
APOSOR937790-10GEA
614
GBP
APOSOR937790-10G
APOSOR937790-1G
APOSOR937790-5G
O-Benzoyl-N-methylhydroxylamine hydrochloride 95%
O-Benzoyl-N-methylhydroxylamine hydrochloride
Formula:
C₈H₉NO₂ Storage Temperature: Ambient |
CAS Number:
27130-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...