1,2-Heptanediol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,2-Dihydroxyheptane

OR937209-10G OR937209-1G OR937209-5G
APOSOR937209-10GEA 494 GBP
APOSOR937209-10G APOSOR937209-1G APOSOR937209-5G
1,2-Heptanediol 95%
1,2-Heptanediol
Formula: C₇H₁₆O₂
MW: 132.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861287
CAS Number: 3710-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR