Hept-6-enoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR937178-1G OR937178-5G
APOSOR937178-1GEA 15.6 GBP
APOSOR937178-1G APOSOR937178-5G
Hept-6-enoic acid 95%
Hept-6-enoic acid
Formula: C₇H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00014382
CAS Number: 1119-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR