Dibenzyl ether 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR925290-100G OR925290-1KG OR925290-500G
APOSOR925290-100GEA 36.9 GBP
APOSOR925290-100G APOSOR925290-1KG APOSOR925290-500G
Dibenzyl ether 95%
Dibenzyl ether
Formula: C₆H₅CH₂OCH₂C₆H₅
Storage Temperature: Ambient
MDL Number: MFCD00004780
CAS Number: 103-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR