N,N-Dimethylisobutyramide 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR925258-25G OR925258-100G
APOSOR925258-25GEA 43.1 GBP
APOSOR925258-25G APOSOR925258-100G
N,N-Dimethylisobutyramide 97%
N,N-Dimethylisobutyramide
Formula: C₆H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00075378
CAS Number: 21678-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR