(R)-(-)-α-Cyclohexanemethylamine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR925101-25G OR925101-5G
APOSOR925101-25GEA 177 GBP
APOSOR925101-25G APOSOR925101-5G
(R)-(-)-α-Cyclohexanemethylamine 95%
(R)-(-)-α-Cyclohexanemethylamine
Formula: C₈H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00043338
CAS Number: 5913-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR