Diethyl sulphate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR924366-100G OR924366-500G
APOSOR924366-100GEA 89.8 GBP
APOSOR924366-100G APOSOR924366-500G
Diethyl sulphate 98%
Diethyl sulphate
Formula: C₄H₁₀O₄S
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS Number: 64-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR