2-Phenoxybenzaldehyde 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR9239-10G OR9239-1G OR9239-25G
APOSOR9239-10GEA 92.3 GBP
APOSOR9239-10G APOSOR9239-1G APOSOR9239-25G
2-Phenoxybenzaldehyde 97%
2-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00800666
CAS Number: 19434-34-5
EINECS: 243-061-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR