4-Hydroxybenzoic acid 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
p-Hydroxybenzoic Acid
OR923629-100G
OR923629-1KG
OR923629-500G
APOSOR923629-100GEA
0
GBP
APOSOR923629-100G
APOSOR923629-1KG
APOSOR923629-500G
4-Hydroxybenzoic acid 95%
4-Hydroxybenzoic acid
Formula:
C₇H₆O₃ Melting Pt: 215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...