Phenylacetaldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR922951-100G OR922951-1KG OR922951-500G OR922951-25G
APOSOR922951-100GEA 89.8 GBP
APOSOR922951-100G APOSOR922951-1KG APOSOR922951-500G APOSOR922951-25G
Phenylacetaldehyde 98%
Phenylacetaldehyde
Formula: C₈H₈O
Boiling Pt: 195 °C
Melting Pt: –10 °C
Density: 1.027 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006993
CAS Number: 122-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR