Cyclohexanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Hydroxycyclohexane, Hexahydrophenol

OR921713-10G OR921713-1G OR921713-5G
APOSOR921713-10GEA 358 GBP
APOSOR921713-10G APOSOR921713-1G APOSOR921713-5G
Cyclohexanol 95%
Cyclohexanol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 161
Melting Pt: 25
Flash Pt: 59
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
Merck Index: 13,02754

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR