5-Hydroxy-4-octanone 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Hydroxyoctan-4-one, 5-Hydroxy-4-octanone
OR920696-25G
OR920696-5G
APOSOR920696-25GEA
88.1
GBP
APOSOR920696-25G
APOSOR920696-5G
5-Hydroxy-4-octanone 95%
5-Hydroxy-4-octanone
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 188 Melting Pt: 2 Storage Temperature: Ambient |
MDL Number:
MFCD00021928 CAS Number: 496-77-5 Merck Index: 13,01595 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...