(-)-Terpinen-4-ol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (-)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (R)-1-Isopropyl-4-methylcyclohex-3-enol, (-)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene, (-)-p-Menth-1-en-4-ol

OR920302-100G
APOSOR920302-100GEA 26.3 GBP
APOSOR920302-100G
(-)-Terpinen-4-ol 95%
(-)-Terpinen-4-ol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Density: 0.934 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00167108
CAS Number: 20126-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR