1-Ethyl-1-phenylhydrazine 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR919806-1G
OR919806-5G
APOSOR919806-1GEA
94.9
GBP
APOSOR919806-1G
APOSOR919806-5G
1-Ethyl-1-phenylhydrazine 95%
1-Ethyl-1-phenylhydrazine
Formula:
C₈H₁₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00031413 CAS Number: 644-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...