5-Chloro[1,2,4]triazolo[4,3-a]pyridine 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chloro-[1,2,4]triazolo[4,3-a]pyridine
OR909064-1G
APOSOR909064-1GEA
57.8
GBP
APOSOR909064-1G
5-Chloro[1,2,4]triazolo[4,3-a]pyridine 95%
5-Chloro[1,2,4]triazolo[4,3-a]pyridine
Formula:
C₆H₄ClN₃ MW: 153.569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109431 CAS Number: 27187-13-9 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...