(S)-(+)-4-Phenyl-2-oxazolidinone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR8745-100G OR8745-25G OR8745-500G
APOSOR8745-100GEA 21.8 GBP
APOSOR8745-100G APOSOR8745-25G APOSOR8745-500G
(S)-(+)-4-Phenyl-2-oxazolidinone 98%
(S)-(+)-4-Phenyl-2-oxazolidinone
Formula: C₉H₉NO₂
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00043396
CAS Number: 99395-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR