4,4'-Dibromobiphenyl (PBB No. 15) 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: PBB No. 15

OR6695-100G OR6695-25G OR6695-500G
APOSOR6695-100GEA 31.6 GBP
APOSOR6695-100G APOSOR6695-25G APOSOR6695-500G
4,4'-Dibromobiphenyl (PBB No. 15) 98%
4,4'-Dibromobiphenyl (PBB No. 15)
Formula: C₁₂H₈Br₂
MW: 312 g/mol
Boiling Pt: 355…360 °C
Melting Pt: 167…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS Number: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR