1,10-Diiododecane 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OR6497-10G
OR6497-250G
OR6497-50G
APOSOR6497-10GEA
52.6
GBP
APOSOR6497-10G
APOSOR6497-250G
APOSOR6497-50G
1,10-Diiododecane 98%
1,10-Diiododecane
Formula:
C₁₀H₂₀I₂ Boiling Pt: 197…200 °C/12mmHg Melting Pt: 33…35 °C Density: 2.35 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001086 CAS Number: 16355-92-3 EINECS: 240-415-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...