2-(Tributylstannyl)pyrazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR6448-1G OR6448-5G
APOSOR6448-1GEA 84.8 GBP
APOSOR6448-1G APOSOR6448-5G
2-(Tributylstannyl)pyrazine
2-(Tributylstannyl)pyrazine

Remainder non-aromatic trialkyl and tetraalkyltins.

Formula: C₁₆H₃₀N₂Sn
Boiling Pt: 130 °C/1mmHg
Density: 1.1706 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD01319077
CAS Number: 205371-27-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR