Hydroxylammonium chloride 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

OR62004-1KG OR62004-5KG
APOSOR62004-1KGEA 52.8 GBP
APOSOR62004-1KG APOSOR62004-5KG
Hydroxylammonium chloride 99%
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 155…157 °C (dec.)
Density: 1.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
UN: 2923
ADR: 8+6.1 III
Merck Index: 13,04853

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR