2-Methoxy-4-(tributylstannyl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR61411-1G OR61411-250MG
APOSOR61411-1GEA 116 GBP
APOSOR61411-1G APOSOR61411-250MG
2-Methoxy-4-(tributylstannyl)pyridine
2-Methoxy-4-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
Storage Temperature: Ambient
MDL Number: MFCD10699153
CAS Number: 1204580-72-2
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR