1,3-Dichloro-2-propanol 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR61346-25G
OR61346-100G
APOSOR61346-25GEA
15.9
GBP
APOSOR61346-25G
APOSOR61346-100G
1,3-Dichloro-2-propanol 95%
1,3-Dichloro-2-propanol
Formula:
ClH₂CCHOHCH₂Cl Boiling Pt: 174…176 °C Melting Pt: 4 °C Density: 1.351 g/cm³ (25 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000951 CAS Number: 96-23-1 UN: 2750 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...