4-[6-(Tributylstannyl)-2-pyridinyl]morpholine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR59449-1G OR59449-250MG
APOSOR59449-1GEA 331 GBP
APOSOR59449-1G APOSOR59449-250MG
4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
Formula: C₂₁H₃₈N₂OSn
Storage Temperature: Ambient
MDL Number: MFCD08271894
CAS Number: 869901-24-6
UN: 2788
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR