Ac-Lys(Boc)-OH 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Nα-Acetyl-Nepsilon-Boc-L-lysine

OR55059-100G OR55059-25G OR55059-5G
APOSOR55059-100GEA 119 GBP
APOSOR55059-100G APOSOR55059-25G APOSOR55059-5G
Ac-Lys(Boc)-OH 95+%
Ac-Lys(Boc)-OH
Formula: C₁₃H₂₄N₂O₅
MW: 288.344 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236761
CAS Number: 23500-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR