Borane-2-methylpyridine complex

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR55051-25G OR55051-5G
APOSOR55051-25GEA 295 GBP
APOSOR55051-25G APOSOR55051-5G
Borane-2-methylpyridine complex
Borane-2-methylpyridine complex
Formula: C₆H₁₀BN
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD07784361
CAS Number: 3999-38-0
UN: 2813
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR