3-Nitrophthalic anhydride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR54546-250G OR54546-100G OR54546-500G
APOSOR54546-250GEA 63.4 GBP
APOSOR54546-250G APOSOR54546-100G APOSOR54546-500G
3-Nitrophthalic anhydride 95%
3-Nitrophthalic anhydride
Formula: C₈H₃NO₅
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005921
CAS Number: 641-70-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR