Methyl isocyanoacetate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR54541-1G OR54541-5G
APOSOR54541-1GEA 68.3 GBP
APOSOR54541-1G APOSOR54541-5G
Methyl isocyanoacetate 95%
Methyl isocyanoacetate
Formula: C₄H₅NO₂
Boiling Pt: 75…76 °C/10mmHg
Density: 1.09 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000006
CAS Number: 39687-95-1
UN: 2922
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR