4,4'-Bis(dimethylamino)benzhydrol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: N,N,N',N'-Tetramethyl-4,4'-diaminodiphenylcarbinol, Michler's Hydrol

OR54540-10G OR54540-1G OR54540-5G
APOSOR54540-10GEA 0 GBP
APOSOR54540-10G APOSOR54540-1G APOSOR54540-5G
4,4'-Bis(dimethylamino)benzhydrol
4,4'-Bis(dimethylamino)benzhydrol
Formula: C₁₇H₂₂N₂O
Melting Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00008313
CAS Number: 119-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR