Benzoyl chloride 99%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OR5454-1KG
OR5454-250G
OR5454-5KG
APOSOR5454-1KGEA
84.8
GBP
APOSOR5454-1KG
APOSOR5454-250G
APOSOR5454-5KG
Benzoyl chloride 99%
Benzoyl chloride
Formula:
C₇H₅ClO Boiling Pt: 198 °C Melting Pt: –1 °C Density: 1.211 g/cm³ (25 °C) Flash Pt: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000653 CAS Number: 98-88-4 EINECS: 202-710-8 UN: 1736 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...