Benzonitrile 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Cyanobenzene, Phenylcyanide

OR54377-1L OR54377-2.5L
APOSOR54377-1LEA 73.6 GBP
APOSOR54377-1L APOSOR54377-2.5L
Benzonitrile 99%
Benzonitrile
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 189…191 °C
Melting Pt: –13 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR